Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCMTD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569557 Supplier Invitrogen™ Supplier No. PA569557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

PCMTD2 gene ontology annotations related to this gene include protein-L-isoaspartate (D-aspartate) O-methyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCMTD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PCMTD2
Gene Accession No. Q8BHD8
Gene Alias 5330414D10Rik; AI314201; C20orf36; chromosome 20 open reading frame 36; PCMTD1; Pcmtd2; protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1; protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2; protein-L-isoaspartate O-methyltransferase domain-containing protein 2; RGD1305684
Gene Symbols PCMTD2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse PCMTD2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245867
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.