Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCNA Antibody (5E6/2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250238IR
Description
PCNA Monoclonal specifically detects PCNA in Yeast samples. It is validated for Western Blot, ELISA.Specifications
PCNA | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PCNA | |
Purified recombinant His6-PCNA from budding yeast, Saccharomyces cerevisiae, produced in Escherichia coli | |
0.1 mL | |
Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Stem Cell Markers | |
5111 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
5E6/2 | |
Western Blot, ELISA | |
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Yeast | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction