Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCNA Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100456JF549
Description
PCNA Polyclonal specifically detects PCNA in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PCNA | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
Rabbit | |
Affinity Purified | |
Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Stem Cell Markers | |
5111 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PCNA | |
The immunogen this antibody was made to, maps to a region between residues 75-125 of human proliferating cell nuclear antigen using the SwissProt entry P12004 (GeneID 5111). | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction