Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCNA Antibody (OTI3D6), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273271AF750
Description
PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PCNA | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
PCNA | |
Full length recombinant protein of human PCNA (NP_002583) produced in E.coli. | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI3D6 | |
Western Blot, Immunohistochemistry | |
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
Mouse | |
0.1 mL | |
5111 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction