Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCNA associated factor Antibody (K11-P1G7*B8), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250536AF405
Description
PCNA associated factor Monoclonal specifically detects PCNA associated factor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
PCNA associated factor | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PCLAF | |
Peptide Sequence - RKACPLQPDHT -peptide immunogen is identical in mouse (Mus musculus) and rat (Rattus norvegicus). | |
0.1 mL | |
Cancer | |
9768 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunohistochemistry | |
K11-P1G7*B8 | |
Western Blot, ELISA, Immunohistochemistry | |
HCV NS5A-transactivated protein 9, Hepatitis C virus NS5A-transactivated protein 9, KIAA0101, NS5ATP9p15PAF, OEATC-1OEATC1, Overexpressed in anaplastic thyroid carcinoma 1, p15(PAF), PAF, PCNA-associated factor | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction