Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCNA associated factor Antibody (K11-P1G7*B8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | PCNA associated factor |
---|---|
Clone | K11-P1G7*B8 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Description
PCNA associated factor Monoclonal antibody specifically detects PCNA associated factor in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
PCNA associated factor | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cancer | |
PBS | |
9768 | |
IgG2b κ | |
Protein A purified |
K11-P1G7*B8 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
HCV NS5A-transactivated protein 9, Hepatitis C virus NS5A-transactivated protein 9, KIAA0101, NS5ATP9p15PAF, OEATC-1OEATC1, Overexpressed in anaplastic thyroid carcinoma 1, p15(PAF), PAF, PCNA-associated factor | |
Peptide Sequence - RKACPLQPDHT -peptide immunogen is identical in mouse (Mus musculus) and rat (Rattus norvegicus). | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title