Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Mouse anti-Human, Mouse, Porcine, Rat, Biotin, Clone: 10D10E11, Proteintech
SDP

Catalog No. 501737225 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7225 Supplier Proteintech Group Inc Supplier No. BIOTIN60097100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein associates with formin-binding protein 17, but its function is unknown. This protein may form oligomeric complexes with family members.

Specifications

Antigen PCNA
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10D10E11
Concentration 0.33 mg/mL
Conjugate Biotin
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PCNA
Gene Accession No. P04961, P12004, P17918
Gene Alias Cyclin, PCNA
Gene Symbols Pcna
Host Species Mouse
Immunogen PCNA Fusion Protein Ag7416
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18538, 25737, 5111, 692192
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.