Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Mouse anti-Human, Mouse, Rat, Clone: 1157CT1.1.3, Abnova™

Catalog No. 89934739 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-739 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-739 Supplier Abnova Corporation Supplier No. MAB12342
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human PCNA.

The protein encoded by this gene is found in the nucleus and is a cofactor of DNA polymerase delta. The encoded protein acts as a homotrimer and helps increase the processivity of leading strand synthesis during DNA replication. In response to DNA damage, this protein is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Two transcript variants encoding the same protein have been found for this gene. Pseudogenes of this gene have been described on chromosome 4 and on the X chromosome. [provided by RefSeq]

Specifications

Antigen PCNA
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1157CT1.1.3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human PCNA.
Dilution Immunohistochemistry (1:25) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias MGC8367
Gene Symbols PCNA
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 236-261 at C-terminus region of human PCNA.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5111
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.