Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCNA, p Tyr211 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10059861
Description
PCNA Polyclonal specifically detects PCNA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PCNA (p Tyr211) | |
Polyclonal | |
Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
P12004 | |
PCNA | |
Immunogen for this antibody was a phosphorylated synthetic peptide, which represented a portion of human proliferating cell nuclear antigen (GeneID 5111) around tyrosine 211 according to the numbering given inentry NP_002583.1. | |
0.1 mL | |
Autophagy, Base Excision Repair, Cell Cycle and Replication, Core ESC Like Genes, DNA Polymerases, DNA Repair, Phospho Specific, Stem Cell Markers | |
5111 | |
Store at 4C. Do not freeze. |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction