Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Rabbit, anti-Human, CoraLite 488, Polyclonal, Proteintech
SDP

Catalog No. 502256755 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-6755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-6755 Supplier Proteintech Group Inc Supplier No. CL48810205100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The PCNA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PCNA. This antibody recognizes human, mouse, rat antigen. The PCNA antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite 488.

Specifications

Antigen PCNA
Applications Flow Cytometry (Intracellular Staining), Immunofluorescence
Concentration 1000 μg/mL
Conjugate CoraLite 488
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3
Host Species Rabbit
Immunogen PCNA fusion protein Ag0277
Purification Method Antigen Affinity Purification
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5111
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.