Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNP Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen PCNP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179669
SDP
View Documents
Novus Biologicals
NBP179669
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

PCNP Polyclonal specifically detects PCNP in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

PCNP
Polyclonal
Rabbit
NP_001019793
57092
Synthetic peptide directed towards the C terminal of human PcnpThe immunogen for this antibody is Pcnp. Peptide sequence AFNEDEDSEPEEMPPEAKMRMKNIGRDTPTSAGPNSFNKGKHGFSDNQKL.
Primary
Western Blot
Unconjugated
RUO
DKFZp781I24156, PEST proteolytic signal containing nuclear protein, PEST proteolytic signal-containing nuclear protein, PEST-containing nuclear protein
PCNP
IgG
20 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.