Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCNP Antibody (OTI4E3), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273272AF405
Description
PCNP Monoclonal specifically detects PCNP in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PCNP | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PCNP | |
Full length recombinant protein of human PCNP (NP_065090) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot | |
OTI4E3 | |
Western Blot | |
DKFZp781I24156, PEST proteolytic signal containing nuclear protein, PEST proteolytic signal-containing nuclear protein, PEST-containing nuclear protein | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
57092.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction