Learn More
Invitrogen™ PCNP Polyclonal Antibody

Description
Immunogen sequence: MADGKAGDEK PEKSQRAGAA GGPEEEAEKP VKTKTVSSSN GGESSSRSAE KRSAEEEAAD LPTKPTKISK FGFAIGSQTT KKASAISIKL GSSKPKETVP TLAPKTLSVA AAFNEDEDSE PEEMPPEAKM RMKNIGRDTP TSAGPNSFNK GKHGFSDNQK LWERNIKSHL GNVHDQDN; Positive Samples: Mouse liver, Rat liver; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | PCNP |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.47 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | PCNP |
| Gene Accession No. | Q6P8I4, Q7TP40 |
| Gene Alias | 1110018D06Rik; Ab2-416; AI647035; Liver regeneration-related protein LRRG084; PCNP; PEST proteolytic signal containing nuclear protein; PEST proteolytic signal-containing nuclear protein; PEST-containing nuclear protein; Unknown (protein for MGC:128940); zgc:103440 |
| Gene Symbols | PCNP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.