Learn More
PCNP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304668AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
PCNP is a protein coding gene, and may be involved in cell cycle regulation.Specifications
PCNP | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8WW12 | |
PCNP | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PCNP | |
PEST proteolytic signal-containing nuclear protein, PEST-containing nuclear protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57092 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.