Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987236
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7236 Supplier Bioss Supplier No. BS6392R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCNP is a protein coding gene, and may be involved in cell cycle regulation.

Specifications

Antigen PCNP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PCNP
Gene Accession No. Q8WW12
Gene Alias 1110018D06Rik; Ab2-416; AI647035; BOS_718; Liver regeneration-related protein LRRG084; LOC100357761; Pcnp; PEST proteolytic signal containing nuclear protein; PEST proteolytic signal-containing nuclear protein; PEST-containing nuclear protein; zgc:103440
Gene Symbols PCNP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PCNP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57092
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.