Learn More
Invitrogen™ PCNX Polyclonal Antibody

Description
Immunogen sequence: VDPSQILEGI NLSKRKELQW PDEGIRLKAG RNSWKDWSPQ EGMEGHVIHR WVPCSRDPGT RSHIDKAVLL VQIDDKYVTV IETGVLELGA EV.
Specifications
Specifications
| Antigen | PCNX |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.04 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | PCNX1 |
| Gene Accession No. | Q96RV3 |
| Gene Alias | KIAA0805; KIAA0995; Pcnx; PCNX1; PCNXL1; pecanex; pecanex 1; pecanex homolog (Drosophila); pecanex homolog 1 (Drosophila); Pecanex homolog protein 1; Pecanex-like protein 1 |
| Gene Symbols | PCNX1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.