Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCNX Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144827 Supplier Invitrogen™ Supplier No. PA5144827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSGRKKECCA GPEEKNSCAS DKRTSSEKIA MEASTNSGVH EAKDPTPSDE MHNQRGLSTS ASEEANKNPH ANEFTSQGDR PPGNTAENKE Antigen sequence identity: Mouse 57%; Rat 56%.

This gene encodes an evolutionarily conserved transmembrane protein similar to the pecanex protein in Drosophila. The fly protein is a component of the Notch signaling pathway, which functions in several developmental processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNX
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCNX1
Gene Accession No. Q96RV3
Gene Alias KIAA0805; KIAA0995; Pcnx; PCNX1; PCNXL1; pecanex; pecanex 1; pecanex homolog (Drosophila); pecanex homolog 1 (Drosophila); Pecanex homolog protein 1; Pecanex-like protein 1
Gene Symbols PCNX1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 426-515. Recombinant protein control fragment (Product #RP-110022).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22990
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.