Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCNX Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565800
Description
Immunogen sequence: TGNNATTANNP HSNVTQGSIG NPGQGSGTGL HPPVTSYPPT LGTSHSSHSV QSGLVRQSPA RASVASQSSY CYSSRHSSLR MSTTGF Highest antigen sequence identity to the following orthologs - mouse 83%, rat 80%.
This gene encodes an evolutionarily conserved transmembrane protein similar to the pecanex protein in Drosophila. The fly protein is a component of the Notch signaling pathway, which functions in several developmental processes.
Specifications
PCNX | |
Polyclonal | |
Unconjugated | |
PCNX1 | |
KIAA0805; KIAA0995; Pcnx; PCNX1; PCNXL1; pecanex; pecanex 1; pecanex homolog (Drosophila); pecanex homolog 1 (Drosophila); Pecanex homolog protein 1; Pecanex-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22990 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96RV3 | |
PCNX1 | |
Recombinant Human PCNX. Recombinant protein control fragment (Product #RP-106472). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction