Learn More
PCNX3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | PCNX3 |
---|---|
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1101
![]() |
Bethyl Laboratories, Inc
A305728AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Pecanex homolog protein 3 (PCNX3) belongs to the pecanex family.Specifications
PCNX3 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
FLJ22427, PCNXL3, pecanex homolog protein 3, pecanex-like 3, pecanex-like 3 (Drosophila) | |
PCNX3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q9H6A9 | |
399909 | |
Between 1984 and 2034 | |
Primary | |
4° C | |
PCNX3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.