Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNXL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501732878 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2878 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2878 Supplier Proteintech Group Inc Supplier No. 204171AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in tumorigenesis of colorectal carcinomas with high microsatellite instability (MSI-H).

Specifications

Antigen PCNXL2
Applications Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PCNX2
Gene Accession No. A6NKB5, Q5DU28
Gene Alias KIAA0435, PCNXL2, pecanex like 2 (Drosophila), Pecanex like protein 2
Gene Symbols Pcnx2
Host Species Rabbit
Immunogen PCNXL2 Fusion Protein Ag14161
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270109, 307949, 80003
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.