Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCNXL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551920
Description
Immunogen sequence: NKSTTVERIL TTDILAEEDE HEFTSCTGAE TVKFLIPGKK YV Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 76%.

Specifications
PCNXL4 | |
Polyclonal | |
Unconjugated | |
PCNX4 | |
1810048J11Rik; 4932413O10; AU017192; C14orf135; FBP2; HCV F protein-binding protein 2; HCV F-interacting protein; hepatitis C virus F protein-binding protein 2; PCNX4; PCNXL4; pecanex homolog 4 (Drosophila); pecanex homolog protein 4; pecanex-like 4; pecanex-like 4 (Drosophila); pecanex-like protein 4; pecanex-like protein C14orf135; pecanex-like protein C14orf135 homolog; RGD1566114 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64430 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q63HM2 | |
PCNX4 | |
Recombinant protein corresponding to Human PCNXL4. Recombinant protein control fragment (Product #RP-89103). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction