Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCNXL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551679 Supplier Invitrogen™ Supplier No. PA551679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELIKYLEEYE RDWYIGLVSD EKWKEAILQE KPYLFSLGYD SNMGIYTGRV LSLQELLIQV GKLNPEAVRG QWANLSWELL YATNDDEERY SIQAHPLLLR NLTVQAAEPP LGYPIYSSKP LHIHLY Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 83%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNXL4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCNX4
Gene Accession No. Q3UVY5, Q63HM2
Gene Alias 1810048J11Rik; 4932413O10; AU017192; C14orf135; FBP2; HCV F protein-binding protein 2; HCV F-interacting protein; hepatitis C virus F protein-binding protein 2; PCNX4; PCNXL4; pecanex homolog 4 (Drosophila); pecanex homolog protein 4; pecanex-like 4; pecanex-like 4 (Drosophila); pecanex-like protein 4; pecanex-like protein C14orf135; pecanex-like protein C14orf135 homolog; RGD1566114
Gene Symbols PCNX4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCNXL4. Recombinant protein control fragment (Product #RP-88904).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64430, 67708, 690217
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.