Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCOLCE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547276
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547276 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547276 Supplier Invitrogen™ Supplier No. PA547276
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant human PCPE is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Fibrillar collagen types I-III are synthesized as precursor molecules known as procollagens. These precursors contain amino- and carboxyl-terminal peptide extensions known as N- and C-propeptides, respectively, which are cleaved, upon secretion of procollagen from the cell, to yield the mature triple helical, highly structured fibrils. This gene encodes a glycoprotein which binds and drives the enzymatic cleavage of type I procollagen and heightens C-proteinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCOLCE
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PCOLCE
Gene Accession No. Q61398
Gene Alias astroglial cell transcript 2; Astt2; Astt-2; P14; Pcolce; PCPE; PCPE1; PCPE-1; procollagen C-endopeptidase enhancer; procollagen C-endopeptidase enhancer 1; procollagen C-endopeptidase enhancer protein; procollagen COOH-terminal proteinase enhancer 1; procollagen C-proteinase enhancer 1; procollagen C-proteinase enhancer protein; procollagen, type 1, COOH-terminal proteinase enhancer; type 1 procollagen C-proteinase enhancer protein; Type I procollagen COOH-terminal proteinase enhancer
Gene Symbols PCOLCE
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PCPE-1 Gln25-Ala468.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18542
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.