Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCOLCE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553034 Supplier Invitrogen™ Supplier No. PA553034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEGKVVVLNF RFIDLESDNL CRYDFVDVYN GHANGQRIGR FCGTFRPGAL VSSGNKMMVQ MISDANTAGN GFMAMFSAAE PNERGDQYCG GLLDRPSGSF KTPNWPDRDY PAGVTCVWHI VAPKNQLIEL KFEKFDVERD NY Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

Binds to the C-terminal propeptide of types I and II procollagens and may enhance the cleavage of that propeptide by BMP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCOLCE2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCOLCE2
Gene Accession No. Q9UKZ9
Gene Alias 2400001O18Rik; PCOLCE2; PCPE2; PCPE-2; Procollagen C-endopeptidase enhancer 2; procollagen COOH-terminal proteinase enhancer 2; procollagen C-proteinase enhancer 2; UNQ250/PRO287
Gene Symbols PCOLCE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCOLCE2. Recombinant protein control fragment (Product #RP-89858).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26577
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.