Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCP4 Monoclonal Antibody (CL5306)
GREENER_CHOICE

Catalog No. PIMA535027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535027 Supplier Invitrogen™ Supplier No. MA535027
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: GAGATNGKDK TSGENDGQKK VQEEFDIDMD APETERAAVA IQSQFRKFQK KK Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.

PCP-4 (Purkinje cell protein 4), also known as PCP4, is a calmodulin (CaM) regulatory protein that is highly expressed in neuronal cells. Through its IQ motif, PCP-4 mediates both the calcium-dependent binding properties of CaM and the rates of association and dissociation of calcium from the C-terminal domain of CaM. The IQ motif contains a serine residue which can be phosphorylated by all isoforms of protein kinase C (PKC). PCP-4 is implicated in uterine leiomyomas, as well as in neurodegenerative disorders such as Alzheimer's disease and Huntington's disease. Additionally, overexpression of PCP-4 is thought to play a role in cerebellar hypoplasia, a key feature of Down syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCP4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL5306
Concentration 0.70 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCP4
Gene Accession No. P48539, P63054, P63055
Gene Alias brain specific polypeptide PEP19; brain-specific antigen PCP-4; Brain-specific polypeptide PEP-19; Calmodulin regulator protein PCP4; P16Rimb19; PCP4; Pcp-4; Pep19; PEP-19; PEPZ19; Purkinje cell protein 4
Gene Symbols PCP4
Host Species Mouse
Immunogen Recombinant protein fragment of Human PCP4 (Product #RP-88803)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18546, 25510, 5121
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.