Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550188 Supplier Invitrogen™ Supplier No. PA550188
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PCP4 protein.

PCP-4 (Purkinje cell protein 4), also known as PCP4, is a calmodulin (CaM) regulatory protein that is highly expressed in neuronal cells. Through its IQ motif, PCP-4 mediates both the calcium-dependent binding properties of CaM and the rates of association and dissociation of calcium from the C-terminal domain of CaM. The IQ motif contains a serine residue which can be phosphorylated by all isoforms of protein kinase C (PKC). PCP-4 is implicated in uterine leiomyomas, as well as in neurodegenerative disorders such as Alzheimer's disease and Huntington's disease. Additionally, overexpression of PCP-4 is thought to play a role in cerebellar hypoplasia, a key feature of Down syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCP4
Applications Western Blot
Classification Polyclonal
Concentration 2.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PCP4
Gene Accession No. P48539, P63054, P63055
Gene Alias brain specific polypeptide PEP19; brain-specific antigen PCP-4; Brain-specific polypeptide PEP-19; Calmodulin regulator protein PCP4; P16Rimb19; PCP4; Pcp-4; Pep19; PEP-19; PEPZ19; Purkinje cell protein 4
Gene Symbols PCP4
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human purkinje cell protein 4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18546, 25510, 5121
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.