Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCP4L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565743
Description
Immunogen sequence: ELNTKTSPATN QAAGQEEKGK AGNVKKAEEE EEIDIDLTAP ETEKAALAIQ GKFR Highest antigen sequence identity to the following orthologs - mouse 85%, rat 85%.
PCP4L1 is a protein coding gene.
Specifications
| PCP4L1 | |
| Polyclonal | |
| Unconjugated | |
| PCP4L1 | |
| 1500036F01Rik; IQM1; PCP4L1; PCP4-like protein 1; Purkinje cell protein 4 like 1; Purkinje cell protein 4-like 1; Purkinje cell protein 4-like protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 654790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NKN8 | |
| PCP4L1 | |
| Recombinant Human PCP4L1. Recombinant protein control fragment (Product #RP-106419). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction