Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ pCPA methyl ester hydrochloride
GREENER_CHOICESDP

Catalog No. 5871100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5871100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5871100 Supplier R&D Systems™ Supplier No. 5871/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Tryptophan hydroxylase inhibitor

pCPA methyl ester hydrochloride is a tryptophan hydroxylase inhibitor. Inhibits 5-HT synthesis. Depletes 5-HT levels in orbitofrontal cortex and impairs reversal learning in rats. Methyl ester of p-Chlorophenylalanine (Cat. No. 0938).

Specifications

Chemical Name or Material 4-Chloro-DL-Phenylalanine methyl ester hydrochloride
CAS 14173-40-1
Quantity 100 mg
Product Line Tocris
Target Hydroxylase Inhibitors
Molecular Formula C10H12ClNO2.HCl
Formula Weight 250.12
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.