Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCPTP1 Monoclonal Antibody (T10), Abbomax

Catalog No. 502204418
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4418 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4418 Supplier Abbomax Inc Supplier No. 606140
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Breast carcinoma Cellular location: Cell membrane and cytoplasm, perinuclear region.

PCPTP1 Monoclonal antibody specifically detects PCPTP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PCPTP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone T10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene PTPRR
Gene Accession No. Q15256
Gene Alias Ch-1 PTPase; ch-1PTPase; ECPPT; ECPTP; EC-PTP; Gmcp1; mPTP213; NC-PTPCOM1; PC12-PTP1; PCPTP1; Phosphotyrosine phosphatase 13; protein tyrosine phosphatase Cr1PTPase; protein tyrosine phosphatase PCPTP1; protein tyrosine phosphatase, receptor type R; protein tyrosine phosphatase, receptor type, R; protein-tyrosine phosphatase NC-PTPCOM1; protein-tyrosine phosphatase PCPTP1; Protein-tyrosine-phosphatase SL; protein-tyrosine-phosphatase-SL; Ptp; Ptp13; PTPBR7; PTPRQ; PTPRR; PTP-SL; receptor-type tyrosine-protein phosphatase R; R-PTP-R; RPTPRR; STEP-like PTPase; tyrosine phosphatase CBPTP
Host Species Mouse
Immunogen A recombinant protein PCPTP1
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.