Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCPTP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567143 Supplier Invitrogen™ Supplier No. PA567143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERFQLSLRQDK EKNQEIHLSP ITLQPALSEA KTVHSMVQPE QAPKVLNVVV DPQGRGAPEI KATTATSVCP SPFK Highest antigen sequence identity to the following orthologs - mouse 80%, rat 80%.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCPTP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPRR
Gene Accession No. Q15256
Gene Alias Ch-1 PTPase; ch-1PTPase; ECPPT; ECPTP; EC-PTP; Gmcp1; mPTP213; NC-PTPCOM1; PC12-PTP1; PCPTP1; Phosphotyrosine phosphatase 13; protein tyrosine phosphatase Cr1PTPase; protein tyrosine phosphatase PCPTP1; protein tyrosine phosphatase, receptor type R; protein tyrosine phosphatase, receptor type, R; protein-tyrosine phosphatase NC-PTPCOM1; protein-tyrosine phosphatase PCPTP1; Protein-tyrosine-phosphatase SL; protein-tyrosine-phosphatase-SL; Ptp; Ptp13; PTPBR7; PTPRQ; PTPRR; PTP-SL; receptor-type tyrosine-protein phosphatase R; R-PTP-R; RPTPRR; STEP-like PTPase; tyrosine phosphatase CBPTP
Gene Symbols PTPRR
Host Species Rabbit
Immunogen Recombinant Human PCPTP1. Recombinant protein control fragment (Product #RP-105518).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.