Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCQAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551848 Supplier Invitrogen™ Supplier No. PA551848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QKLVSQIEDA MRKAGVAHSK SSKDMESHVF LKAKTRDEYL SLVARLIIHF RDIHNKKSQA SVSDPMNALQ SLTGGPAAGA AGIGMPPRGP GQSLGGMGSL GAMGQPMSLS GQP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene encodes a transcription factor that represses GATA-regulated genes and binds to a dynein light chain protein. Binding of the encoded protein to the dynein light chain protein affects binding to GATA consensus sequences and suppresses its transcriptional activity. Defects in this gene are a cause of tricho-rhino-phalangeal syndrome (TRPS) types I-III.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCQAP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MED15
Gene Accession No. Q96RN5
Gene Alias A230074L19Rik; Activator-recruited cofactor 105 kDa component; activator-recruited cofactor, 105-kD; ARC105; AW536074; CAG7A; CTG repeat protein 7a; CTG7A; DKFZp686A2214; DKFZp762B1216; FLJ42282; FLJ42935; MED15; Mediator complex subunit 15; mediator of RNA polymerase II transcription subunit 15; mPcqap; PC2 (positive cofactor 2, multiprotein complex) glutamine/Q-rich-associated protein; PC2 glutamine/Q-rich-associated protein; PC2-glutamine-rich-associated protein; PCQAP; Positive cofactor 2 glutamine/Q-rich-associated protein; positive cofactor 2, glutamine/Q-rich-associated protein; positive cofactor 2, multiprotein complex, glutamine/Q-rich-associated protein; TIG1; TIG-1; TNRC7; TPA inducible gene-1; TPA inducible protein; TPA-inducible gene 1 protein; trinucleotide repeat containing 7; trinucleotide repeat-containing gene 7 protein
Gene Symbols MED15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCQAP. Recombinant protein control fragment (Product #RP-91488).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51586
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.