Learn More
G-Biosciences PCR Clean Up Magnetic Beads
SCI_ED
Catalog No. S04205
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04205 5 mL
S04220 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04205 Supplier G-Biosciences Supplier No. 7862161
Only null left
Add to Cart
Add to Cart
PCR Clean up Magnetic beads are suitable for PCR product clean-up 150 bp to 1000 bp and even larger.
PCR clean up Magnetic Beads remove excess nucleotides, salts, enzymes, and other impurities with a simple washing process to obtain purified fragments, which can be directly used in downstream applications such as sequencing, hybridization, and enzyme digestion. PCR clean up depends on optimizing magnetic beads, target DNA ratio, and salt concentrations.
Specifications
Product Type Magnetic Beads
Primer Random
Quantity 5 mL