Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCR Markers, Thermo Scientific Chemicals

Catalog No. aaj76710amm
Click to view available options
Quantity:
250 μL

Includes

6X DNA Loading Buffer (OXG) with orange G and xylene cyanol FF

 

  • Mixture of 8 DNA bands ranging in size from 50 to 2000 bp
  • Recommended amount for loading is 5μL per gel lane
  • 6X DNA Loading Buffer (OXG) with orange G and xylene cyanol FF is included for convenience to prepare PCR samples for agarose gel electrophoresis
  • Tracking dyes do not interfere with UV illumination of ethidium bromide stained gels
  • Fragments: 2000 bp; 1500 bp; 1000 bp; 750 bp; 500 bp; 300 bp; 150 bp; 50 bp
TRUSTED_SUSTAINABILITY

Specifications

Buffer 6 x DNA Loading Buffer (OXG)
Color Blue, Orange
Concentration 6 x Loading Dye
Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Quantity 250 μL
Storage Requirements 4°C for short-term use, or -20°C for long-term storage.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.