Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ PCR Nucleotide Mix

Catalog No. p-106398 Shop All Fisherbrand Products
Click to view available options
Quantity:
200 μL

A premixed 10mM solution of dATP, dCTP, dGTP, and dTTP used for PCR

  • Functionally tested in PCR
  • High purity: >98% triphosphate
  • Supplied as 10mM solution in purified water (pH 7.5)

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7.5
Content And Storage -20°C
Concentration 10 mM
Purity >98%
Quantity 200 μL
Color Colorless
For Use With (Application) PCR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.