Learn More
Promega PCR Nucleotide Mix, 25mM, 1,000microl
Catalog No. PRU1432 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1,000microl
Catalog No. Quantity
PRU1432 1,000microl
1 options
This item is not returnable. View return policy
Catalog No. PRU1432 Supplier Promega Supplier No. U1432
This item is not returnable. View return policy
PCR Nucleotide Mix is a premixed solution containing the sodium salts of dATP, dCTP, dGTP and dTTP, each at a concentration of 10mM or 25mM in water.
  • cGMP−Grade Solution of dNTPs Pretested in PCR
  • cGMP−manufactured for lot−to−lot product consistency
  • Equimolar amounts of each dNTP in water
  • Two concentrations available: 10mM and 25mM
Specifications
Content And Storage -30C TO -10C
Quantity 1,000microl