Learn More
Greiner Bio-One PCR Strip Tubes without Cap
Catalog No. 07000311 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Assorted
Blue
Green
Natural
Violet
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-311 Assorted
07-000-708 Blue
07-000-709 Green
07-000-711 Violet
07-000-310 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-000-311 Supplier Greiner Bio-One Supplier No. 673271
Add to Cart
Add to Cart
For use in research. Greiner Bio-One PCR Strip Tubes without Cap are free of detectable DNase, Rnase and human DNA.
  • Non-Pyrogenic
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
TRUSTED_SUSTAINABILITY
Specifications
Color Assorted