Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One PCR Strip Tubes without Cap

Catalog No. p-7075434 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Assorted
Blue
Green
Natural
Violet
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-708 Blue
07-000-709 Green
07-000-711 Violet
07-000-311 Assorted
07-000-310 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-000-708 Supplier Greiner Bio-One Supplier No. 673274
Add to Cart
Add to Cart

For use in research. Greiner Bio-One PCR Strip Tubes without Cap are free of detectable DNase, Rnase and human DNA.

  • Non-Pyrogenic
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Volume (Metric) 0.2 mL
For Use With (Equipment) PCR
Sterility Non-sterile
Material Polypropylene
Product Type PCR Tube Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.