Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PCR Tubes and Plates

Catalog No. 2140297
Change view
Click to view available options
Color:
Green
Natural
Red
Yellow
Closure Type:
Domed Cap
Flat Cap
Volume (Metric):
0.2 mL
0.5 mL
Product Type:
PCR Tube
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type Volume (Metric) Product Type
21-402-97 Yellow Flat Cap 0.5 mL PCR Tube
21-236-66 Natural Domed Cap 0.2 mL PCR Tube
21-402-96 Green Flat Cap 0.5 mL PCR Tube
21-402-97A Red Flat Cap 0.5 mL PCR Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-402-97 Supplier Thermo Scientific™ Supplier No. 3430Y

Efficiently perform applications with these PCR tubes and plates, molded from 100% virgin polypropylene.

Efficiently perform applications with Thermo Scientific™ PCR Tubes and Plates, molded from 100% virgin polypropylene.

  • Tubes feature thin walls for optimal heat transfer
  • Caps seal securely to minimize evaporation.
  • Elevated well rims to ensure uniform sealing.
  • Thermo Scientific™ PurePak™ packaging reduces contamination
  • Certified RNase-, DNase-, DNA, and Pyrogen-free
  • Clear, nonsterile
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Volume (Metric) 0.5 mL
For Use With (Equipment) Thermal Cyclers
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Closure Included Yes
Product Type PCR Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.