Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCSK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA120317
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA120317 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA120317 Supplier Invitrogen™ Supplier No. PA120317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-20317 detects PC1 in human, rat and non-human primate samples.

The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a type I proinsulin-processing enzyme that plays a key role in regulating insulin biosynthesis. It is also known to cleave proopiomelanocortin, prorenin, proenkephalin, prodynorphin, prosomatostatin and progastrin. Mutations in this gene are thought to cause obesity. This encoded protein is associated with carcinoid tumors. The use of alternate polyadenylation sites has been found for this gene. Multiple alternatively spliced transcript variants have been described for this gene but their full length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCSK1
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.09% sodium azide
Gene PCSK1
Gene Accession No. P28840, P29120
Gene Alias Att-1; Bdp; BMIQ12; EC 3.4.21.93; Furin homolog; LOW QUALITY PROTEIN: neuroendocrine convertase 1; NEC 1; NEC1; Nec-1; Neuroendocrine convertase 1; PC1; PC3; PCSK1; Phpp-1; Prohormone convertase 1; prohormone convertase 1/3; prohormone convertase 3; propeptide-processing protease; Proprotein convertase 1; proprotein convertase subtilisin/kexin type 1; Protein convertase subtilisin / kexin type I; Protein convertase subtilisin / kexin, type I; SPC3
Gene Symbols PCSK1
Host Species Rabbit
Immunogen Recombinant, full length, rabbit PC1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25204, 5122
Target Species Human, Rat, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.