Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCSK1N Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24942325UL
Description
PCSK1N Polyclonal antibody specifically detects PCSK1N in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
PCSK1N | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
granin-like neuroendocrine peptide, Proprotein convertase 1 inhibitor, proprotein convertase subtilisin/kexin type 1 inhibitorPROSAAS, proSAAS, pro-SAAS, SAAS | |
This antibody was developed against a recombinant protein corresponding to amino acids: PVYDDGPAGPDAEEAGDETPDVDPELLRYLLGRILAGSADSE | |
25 μL | |
metabolism | |
27344 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction