Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCSK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542378 Supplier Invitrogen™ Supplier No. PA542378
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CAGAGADGCI NCTEGYFMED GRCVQSCSIS YYFDHSSENG YKSCKKCDIS Sequence homology: Cow: 100%; Dog: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 79%.

PCSK5 belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. PCSK5 mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known. The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCSK5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Pcsk5
Gene Accession No. Q92824
Gene Alias b2b1549Clo; b2b585Clo; PC5; PC5/6A; PC5A; PC6; PC6A; PCSK5; prohormone convertase 5; proprotein convertase 5; Proprotein convertase 6; proprotein convertase PC5; proprotein convertase subtilisin/kexin type 5; protease PC6; rPC5; SPC6; subtilase; Subtilisin/kexin-like protease PC5; subtilisin-like proprotein convertase 6
Gene Symbols Pcsk5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PCSK5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5125
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.