Learn More
Invitrogen™ PCSK6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532966
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Rat, Bovine, Elephant, Horse, Rabbit (100%); Gibbon, Mouse, Dog, Panda, Pig (93%); Turkey, Chicken (86%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment with Proteinase K.
The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a calcium-dependent serine endoprotease that can cleave precursor protein at their paired basic amino acid processing sites. Some of its substrates are - transforming growth factor beta related proteins, proalbumin, and von Willebrand factor. This gene is thought to play a role in tumor progression. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| PCSK6 | |
| Polyclonal | |
| Unconjugated | |
| PCSK6 | |
| C86343; im:7140527; kexin-like protease PC7A; PACE4; Paired basic amino acid cleaving enzyme 4; paired basic amino acid cleaving system 4; Pcsk6; Proprotein convertase subtilisin/kexin type 6; SPC4; Subtilisin - like endoprotease; Subtilisin/kexin-like protease PACE4; subtilisin-like proprotein convertase 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100057677, 100347173, 25507, 5046, 524684 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P29122, Q63415 | |
| PCSK6 | |
| Synthetic 14 amino acid peptide from Cys-rich motif (CRM) region of human PACE4. | |
| 50 μg | |
| Primary | |
| Human, Rat, Rabbit, Monkey, Bovine, Horse | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.