Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCSK6 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142420
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize reported isoforms a, b, g, h (NP_002561.1; NP_612192.1; NP_612193.1; NP_612194.1 resp.). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a calcium-dependent serine endoprotease that can cleave precursor protein at their paired basic amino acid processing sites. Some of its substrates are - transforming growth factor beta related proteins, proalbumin, and von Willebrand factor. This gene is thought to play a role in tumor progression. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
PCSK6 | |
Polyclonal | |
Unconjugated | |
PCSK6 | |
C86343; im:7140527; kexin-like protease PC7A; PACE4; Paired basic amino acid cleaving enzyme 4; paired basic amino acid cleaving system 4; Pcsk6; Proprotein convertase subtilisin/kexin type 6; SPC4; Subtilisin - like endoprotease; Subtilisin/kexin-like protease PACE4; subtilisin-like proprotein convertase 4 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5046 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P29122 | |
PCSK6 | |
Peptide with sequence PDCEPGTYFDSE. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction