Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCSK7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571875
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571875 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571875 Supplier Invitrogen™ Supplier No. PA571875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCSK7 Polyclonal Antibody for Western Blot

PCSK7 is likely to represent a ubiquitous endoprotease activity within constitutive secretory pathways and capable of cleavage at the RXXX[KR]R consensus motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCSK7
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Pcsk7
Gene Accession No. Q16549
Gene Alias AA959856; hPC8; LPC; Lymphoma proprotein convertase; Pc7; PC8; PCSK7; Prohormone convertase 7; prohormone convertase PC7; proprotein convertase 7; Proprotein convertase 8; proprotein convertase PC7; proprotein convertase subtilisin/kexin type 7; proprotein convertase subtilisin/kexin type 7 precursor variant 2; rPC7; SPC7; subtilisin/kexin-like protease PC7; Subtilisin-like proprotein convertase 7
Gene Symbols Pcsk7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 692-721 amino acids from the C-terminal region of human PCSK7.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9159
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.