Learn More
Invitrogen™ PCSK9 Monoclonal Antibody (407119)

Description
This antibody detects the mature form of human and mouse Proprotein Convertase 9/PCSK9 in direct ELISAs and Western blots. This antibody does not cross-react with recombinant human (rh) PC-1 or rhPC-7. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | PCSK9 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 407119 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | PCSK9 |
| Gene Accession No. | Q80W65, Q8NBP7 |
| Gene Alias | AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.