Learn More
Invitrogen™ PCSK9 Monoclonal Antibody (499911)

Description
In Western blots, 10% cross-reactivity with recombinant mouse PCSK9 is observed and no cross-reactivity with recombinant human (rh) PCSK1 or rhPCSK7 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | PCSK9 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 499911 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | PCSK9 |
| Gene Accession No. | Q8NBP7 |
| Gene Alias | AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.