Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCSK9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596836
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596836 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596836 Supplier Invitrogen™ Supplier No. PA596836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SW480; Cellular Location: Cell surface, Cytoplasm, Endoplasmic reticulum, Endosome, Golgi apparatus, Lysosome.

PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCSK9
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PCSK9
Gene Accession No. P59996, Q80W65, Q8NBP7
Gene Alias AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9
Gene Symbols PCSK9
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100102, 255738, 298296
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.