Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCSK9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA580885
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PCSK9/NARC1.
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
Specifications
PCSK9 | |
Polyclonal | |
PBS with no preservative | |
Q8NBP7 | |
PCSK9 | |
Recombit Human PCSK9/NARC1 protein | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PCSK9 | |
Proprotein convertase subtilisin/kexin type 9; Proprotein convertase PC9; Subtilisin/kexin-like protease PC9; Neural apoptosis-regulated convertase 1; NARC-1; PSEC0052; NARC1; PCSK9 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
255738 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction