Learn More
PCSK9 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305107AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Monkey, Gorilla and Chimpanzee
This gene encodes a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. The encoded protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. This protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons. Mutations in this gene have been associated with a third form of autosomal domit familial hypercholesterolemia .Specifications
| PCSK9 | |
| Polyclonal | |
| Unconjugated | |
| PCSK9 | |
| Proprotein convertase subtilisin/kexin type 9; Proprotein convertase PC9; Subtilisin/kexin-like protease PC9; Neural apoptosis-regulated convertase 1; NARC-1; PSEC0052; NARC1; PCSK9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 255738 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8NBP7 | |
| PCSK9 | |
| Between 450 and 500 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.