Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCTAIRE1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232254AF700
Description
PCTAIRE1 Polyclonal antibody specifically detects PCTAIRE1 in Human samples. It is validated for Western BlotSpecifications
PCTAIRE1 | |
Polyclonal | |
Western Blot | |
Cell division protein kinase 16, cyclin-dependent kinase 16, EC 2.7.11, EC 2.7.11.22, FLJ16665, PCTAIRE, PCTAIRE protein kinase 1, PCTAIRE1, PCTAIRE-motif protein kinase 1, PCTGAIRE, PCTK1Serine/threonine-protein kinase PCTAIRE-1 | |
The immunogen this antibody was made to, maps to a region between residue 25 to 75 of human PCTAIRE-Motif Protein Kinase 1 using the numbering given in entry NP_006192.1 (GeneID 5127). | |
0.1 mL | |
Cell Cycle and Replication | |
5127 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction